Order Entry
Poland
Orders LinkContactUsLinkComponent
Bottle top tilt measures
Bottle top tilt measures
40 ml
Catalog # 613-8719
Supplier:   witeg Labortechnik
Bottle top tilt measures
Catalog # 613-8719
Supplier:  witeg Labortechnik
Bottle top tilt measures
Bottle top tilt measures
40 ml
Price
300 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
Glass, with normal-shaped core.
  • For repetitive dispensing of the same volume
  • Can bleed with all bottles with NS 29/32
Specifications
  • Capacity:
    40 ml