Order Entry
Poland
Orders LinkContactUsLinkComponent
HPLC columns, MetaSil® Basic
HPLC columns, MetaSil® Basic
Guard column, 100 Å, 3 µm
Catalog # VARIA0516MG2
Supplier:   VARIAN
HPLC columns, MetaSil® Basic
Catalog # VARIA0516MG2
Supplier:  VARIAN
HPLC columns, MetaSil® Basic
Guard column, 100 Å, 3 µm
Price
Inquire For Price
Log in for availability
1 * 3 SZT
Product Details & Documents
MetaSil® Basic columns tends to retain polar compounds better than standard phases, often showing improved retention for compounds that elute in the void volume on standard alkyl phases. These columns are recommended for use at pH 2 to 7,5.
  • Excellent peak shape for basic analytes
  • Exceptionally high surface coverage
  • Improved polar compound retention
  • Enhanced selectivity
Specifications
  • Column Format:
    Guard column
  • Particle Size:
    3 µm
  • Pore size:
    100 Å
  • Seq:
    04