(±)-1-Methoxy-2-propanol ≥99%
Glass bottle, 1.000 mL
Supplier: Thermo Fisher Scientific
(±)-1-Methoxy-2-propanol ≥99%
Catalog # L12694.AU
Supplier: Thermo Fisher Scientific
CAS Number: 107-98-2
Jump to:
Product Details & Documents
Synonyms: (±)-Monopropylene glycol methyl ether, 1-Methoxy-2-propanol
- CAS Number:107-98-2
- Formula:CH₃CH(OH)CH₂OCH₃
- MDL Number:MFCD00004537
- Molecular Weight:90.12 g/mol
- Boiling Pt:119.6 °C (1013 hPa)
- Melting Pt:–97 °C
- Density:0.92 g/cm³ (20 °C)
- Flash Pt:31 °C
- Storage Temperature:Ambient
- EINECS:203-539-1
- UN:3092
- ADR:3,III
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL



