Order Entry
Poland
Orders LinkContactUsLinkComponent
Microplates, opaque OptiPlates™
Microplates, opaque OptiPlates™
OptiiPlate™-96 microplates, 96, White, Case of 200
Catalog # PERK6005299
Marketsource
Supplier:   PerkinElmer
Microplates, opaque OptiPlates™
Catalog # PERK6005299
Supplier:  PerkinElmer
Marketsource
Microplates, opaque OptiPlates™
OptiiPlate™-96 microplates, 96, White, Case of 200
Price
Inquire For Price
Log in for availability
1 * 1 SZT
Product Details & Documents
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
  • Made of polystyrene
Specifications
  • No. of wells:
    96
  • Packed:
    Case of 200
  • Colour:
    White
  • Description:
    OptiiPlate™-96 microplates
  • Seq:
    03