Order Entry
Poland
Orders LinkContactUsLinkComponent
Powder funnels, Quickfit®
Powder funnels, Quickfit®
24/29 NS, 105 mm
Catalog # 201-8296
Supplier:   SciLabware
Powder funnels, Quickfit®
Catalog # 201-8296
Supplier:  SciLabware
Powder funnels, Quickfit®
Powder funnels, Quickfit®
24/29 NS, 105 mm
Price
189 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
PYREX®, borosilicate glass, stem with cone.
  • Very good chemical resistance
  • High temperature resistance
Designed for transfer of powders into flasks and vessels.
Specifications
  • Cone size:
    24/29 NS
  • Funnel O-Ø:
    105 mm
  • Seq:
    03