Benzaldehyde ≥98%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 100-52-7 Molecular Formula: C7H6O Formula Weight: 106.12 Flash Point: 62°(143°F) Physical Form: Liquid Density: 1.047 Refractive Index: 1.5450 Sensitivity: Air Sensitive Appearance: Colourless Odour: Almond-like Melting Point: -26° Boiling Point: 178-179° Beilstein Reference: 471223 Merck Reference: 14,1058 UN No.: 1990 MDL No.: MFCD00003299
- CAS Number:100-52-7
- Formula:C₆H₅CHO
- MDL Number:MFCD00003299
- Molecular Weight:106.12 g/mol
- Merck Index:14,1058
- Boiling Pt:178…179 °C
- Melting Pt:–26 °C
- Density:1.047
- Flash Pt:62 °C (143 °F)
- Storage Temperature:Ambient
- EINECS:202-860-4
- UN:1990
- ADR:9,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



