D-Amygdalin hydrate 96%
2 g
Supplier: Thermo Fisher Scientific
D-Amygdalin hydrate 96%
Catalog # L12731.04
Supplier: Thermo Fisher Scientific
CAS Number: 29883-15-6
Jump to:
Product Details & Documents
Purity: 96% CAS No.: 29883-15-6 Molecular Formula: C20H27NO11.xH2O Formula Weight: 457.44(anhy) Melting Point: 215-220° Beilstein Reference: 66856 Optical Rotation: -36° (c=1.2 in water) MDL No.: MFCD00006598
Synonyms: D-Mandelonitrile-beta-D-glucosido-6-beta-D-glucoside hydrate, Amygdalin hydrate
- CAS Number:29883-15-6
- Formula:C₂₀H₂₇NO₁₁·xH₂O
- MDL Number:MFCD00006598
- Molecular Weight:457.44 g/mol (anhy)
- Merck Index:14,597
- Melting Pt:215…220 °C
- Storage Temperature:Ambient
- EINECS:249-925-3
Specifications
Specification Test Results
- Appearance (Color):White
- Form:Crystalline solid
- Assay (HPLC):>95.0%
- Loss on Drying:<10.0% @ 110°C
- Optical Rotation:-38 ±2° (c=1.2 in water) on dry wt. basis
Specifications
- Pk:2 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



