Order Entry
Poland
Orders LinkContactUsLinkComponent
1,4-Butanediol diglycidyl ether 96%

1,4-Butanediol diglycidyl ether 96%

Glass bottle for solids, 50 g
Catalog # L12632.18
Irritant
1,4-Butanediol diglycidyl ether 96%
Catalog # L12632.18
CAS Number:  2425-79-8
1,4-Butanediol diglycidyl ether 96%
1,4-Butanediol diglycidyl ether 96%

Glass bottle for solids, 50 g

CAS Number:  2425-79-8
Price
1 310 zł
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 96% CAS No.: 2425-79-8 Molecular Formula: C10H18O4 Formula Weight: 202.25 Flash Point: 140°(284°F) Refractive Index: 1.4550 Boiling Point: 157-158°/11mm Beilstein Reference: 115238 MDL No.: MFCD00005146

Synonyms:  Araldite RD-2, 1,4-Bis(oxiran-2-ylmethoxy)butane
  • CAS Number:2425-79-8
  • Formula:C₁₀H₁₈O₄
  • MDL Number:MFCD00005146
  • Molecular Weight:202.25 g/mol
  • Boiling Pt:157…158 °C (11 mmHg)
  • Density:1.049
  • Flash Pt:140 °C (284 °F)
  • Storage Temperature:Ambient
  • EINECS:219-371-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.