Order Entry
Poland
Orders LinkContactUsLinkComponent
(±)-1,2-Octanediol ≥97%

(±)-1,2-Octanediol ≥97%

Glass bottle for solids, 50 g
Catalog # L08031.18
Irritant
(±)-1,2-Octanediol ≥97%
Catalog # L08031.18
CAS Number:  1117-86-8
(±)-1,2-Octanediol ≥97%
(±)-1,2-Octanediol ≥97%

Glass bottle for solids, 50 g

CAS Number:  1117-86-8
Price
811 zł
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 97% CAS No.: 1117-86-8 Molecular Formula: C8H18O2 Formula Weight: 146.23 Flash Point: 138°(280°F) Melting Point: 28-34° Boiling Point: 131-133°/10mm Beilstein Reference: 1719619 MDL No.: MFCD00010738

Synonyms:  1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
  • CAS Number:1117-86-8
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00010738
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:131…133 °C (10 mmHg)
  • Melting Pt:28…34 °C
  • Flash Pt:138 °C (280 °F)
  • Storage Temperature:Ambient
  • EINECS:214-254-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications