Order Entry
Poland
Orders LinkContactUsLinkComponent
Bromophenol blue in aqueous solution

Bromophenol blue in aqueous solution

Plastic bottle, 25 mL
Catalog # LOVI451620
Supplier:   Lovibond
Bromophenol blue in aqueous solution
Catalog # LOVI451620
Supplier:  Lovibond
CAS Number:  115-39-9
Bromophenol blue in aqueous solution

Plastic bottle, 25 mL

CAS Number:  115-39-9
Price
102 zł
Log in for availability
1 * 25 mL
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mL

Specifications