Order Entry
Poland
Orders LinkContactUsLinkComponent
Calcium standard solution

Calcium standard solution

Calcium, Ca, 1000 mg/L, H₂O, 1 L
Catalog # HACH12153
Supplier:   Hach
Calcium standard solution
Catalog # HACH12153
Supplier:  Hach
CAS Number:  
Calcium standard solution
Calcium standard solution

Calcium, Ca, 1000 mg/L, H₂O, 1 L

CAS Number:  
Price
343 zł
Log in for availability
1 * 1 L
Product Details & Documents

Standard solution for accuracy checks in calcium hardness determinations.

Documents

Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    1 L
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.