Order Entry
Poland
ContactUsLinkComponent
Thermometer, contact, with rotary setting magnet, mercury filling, measuring range:0:100/1C, standard cap, milk glass scale, capill ary yellow backed, upper part 245x18-19mm, 50mm immersion. 1 * 1 items
  AMARS561126
 :  Amarell
Thermometer, contact, with rotary setting magnet, mercury filling, measuring range:0:100/1C, standard cap, milk glass scale, capill ary yellow backed, upper part 245x18-19mm, 50mm immersion. 1 * 1 items
  AMARS561126
 :  Amarell
Frequently Bought Together