Order Entry
Poland
Orders LinkContactUsLinkComponent
Molecular models of elements, Molymod®
Nitrogen, Blue, 4, 23 mm
Catalog # SPIRMA-401
Marketsource
Supplier:   SPIRING ENTERPRISES
Molecular models of elements, Molymod®
Catalog # SPIRMA-401
Marketsource
Molecular models of elements, Molymod®
Molecular models of elements, Molymod®
Nitrogen, Blue, 4, 23 mm
Price
Inquire For Price
Log in for availability
1 * 10 SZT
Product Details & Documents
Molymod is the original, unique, dual-scale system of high quality molecular models.
Specifications
  • Colour:
    Blue
  • Description:
    Nitrogen
  • No. of holes:
    4
  • Ø:
    23 mm