ACETYLCYSTEINE IMP. B (EP): L-CYSTEINE 1 * 100 mg
Catalog # PROOMM0024.02
Supplier: LGC Standards PROMOCHEM
ACETYLCYSTEINE IMP. B (EP): L-CYSTEINE 1 * 100 mg
Catalog # PROOMM0024.02
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Toluene
Supplier: CHEMPUR
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
ETHANOL 80% SOLUTION 1 * 500 mL
Supplier: CHEMPUR
ETHYL ACETATE EXTRA PURE, DAC, N. F. 1 * 100 L
Supplier: Honeywell Chemicals
Ethylcellulose, Reference Standard
Supplier: USP
2-PROPANOL CZDA OPAK. Z TW. 1 * 5 L
Supplier: CHEMPUR
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND