TRAZODONE HYDROCHLORIDE IMPURITY STANDAR 1 * 50 mg
Catalog # PROOBP591
Supplier: LGC Standards PROMOCHEM
TRAZODONE HYDROCHLORIDE IMPURITY STANDAR 1 * 50 mg
Catalog # PROOBP591
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Acetonitrile, Prepsolv® for preparative chromatography
Supplier: Merck
Chloroform analytical reagent
Supplier: POCH AVANTOR BRAND
Carbon disulphide ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS for analysis
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
Chlorobenzene, Sigma-Aldrich®
Supplier: Merck