Order Entry
Poland
Orders LinkContactUsLinkComponent
DNASE 1 BUFFER (10X) 1 * 1 ml

DNASE 1 BUFFER (10X) 1 * 1 ml

Catalog # AMBI8170G
Marketsource
Supplier:   Ambion
DNASE 1 BUFFER (10X) 1 * 1 ml
Catalog # AMBI8170G
Supplier:  Ambion
Marketsource
DNASE 1 BUFFER (10X) 1 * 1 ml
Price
Inquire For Price
Log in for availability
1 * 1 mL
Product Details & Documents