Order Entry
Poland
Orders LinkContactUsLinkComponent
LC/MS calibrant mixes

LC/MS calibrant mixes

ES-TOF tuning mix, 100 ml, 100 mL
Catalog # HEWLG1969-85000
Supplier:   Agilent
LC/MS calibrant mixes
Catalog # HEWLG1969-85000
Supplier:  Agilent
CAS Number:  
LC/MS calibrant mixes
LC/MS calibrant mixes

ES-TOF tuning mix, 100 ml, 100 mL

CAS Number:  
Price
1 390 zł
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    ES-TOF tuning mix, 100 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.