Blender Bags
Standard 400: quality bag designed to fit any 400 ml paddle blender machines, manufactured from extruded polymer tube using high-grade, food quality polythene, 10 sachets of 50 bags, 65 µm, 180×300 mm
Dostawca: Grade Products
Przejdź do:
Product Details & Documents
High grade polymer mixture, ideal for any food laboratory sample blenders.
- With write-on panel to allow for easy identification of samples, useful for barcoding
Sterilisation by 15 to 40 kGy.
The Lab 400 Blender Bag is manufactured from a 21st century polymer which gives the bag the same strength as a Standard 400 bag, whilst being over 20% thinner. It is ideal for high sampling food testing laboratories.
Documents
Certyfikaty
Szukaj według numeru partiiDane techniczne
- Packed:10 sachets of 50 bags
- Description:Standard 400: quality bag designed to fit any 400 ml paddle blender machines, manufactured from extruded polymer tube using high-grade, food quality polythene
- Seq:02
- Thickness:65 µm
- W×L:180×300 mm



