TLC polyester sheets
POLYGRAM® TLC polyester sheets CEL 300, Cellulose, 40×80 mm
Catalog # 552-0279
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Documents
Thin Layer Chromatography.pdf
Additional Product Details
Specifications
- Coating material:Cellulose
- Plate size:40×80 mm
- Description:POLYGRAM® TLC polyester sheets CEL 300
- Seq:03
- Fluorescent indicator:No