Order Entry
Poland
Orders LinkContactUsLinkComponent
L(-)-Proline

L(-)-Proline

Glass bottle for solids, 100 g
Catalog # A1707.0100
Supplier:   PanReac AppliChem
L(-)-Proline
Catalog # A1707.0100
Supplier:  PanReac AppliChem
CAS Number:  147-85-3
L(-)-Proline

Glass bottle for solids, 100 g

CAS Number:  147-85-3
Price
283 zł
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
  • CAS Number:147-85-3
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00064318
  • Molecular Weight:115.13 g/mol
  • Merck Index:13,07871
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:220…222 °C
  • Density:1.35 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:205-702-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications