Order Entry
Poland
Orders LinkContactUsLinkComponent
Base materials

Base materials

Peptone from casein (pancreatic digest), Glass bottle for solids, 500 g
Catalog # APPCA2210.0500
Supplier:   PanReac AppliChem
Base materials
Catalog # APPCA2210.0500
Supplier:  PanReac AppliChem
CAS Number:  
Base materials
Base materials

Peptone from casein (pancreatic digest), Glass bottle for solids, 500 g

CAS Number:  
Price
439 zł
Log in for availability
1 * 500 g
Product Details & Documents

Components of many microbiological growth media.

  • Store in a dry place
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Description:
    Peptone from casein (pancreatic digest)

Specifications