Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Gentamicin sulphate

Glass bottle for solids, 25 g
Catalog # A1492.0025
Health
Supplier:   PanReac AppliChem
Gentamicin sulphate
Catalog # A1492.0025
Supplier:  PanReac AppliChem
CAS Number:  1405-41-0
Gentamicin sulphate

Glass bottle for solids, 25 g

CAS Number:  1405-41-0
Price
529 zł
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Merck Index:13,04403
  • Melting Pt:218…237 °C
  • Storage Temperature:Ambient
  • EINECS:215-778-9

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications