SnapRing Vial 1.5ml,32x11.6mm, label 1 * 100 items
Catalog # 548-3089
Supplier: LA PHA PACK
Frequently Bought Together
Glass snap top vials, 2 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Crimp caps for crimp neck vials, N 20
Supplier: MACHEREY-NAGEL
HPLC columns, LiChrospher®
Supplier: Merck
VWR® ND9, Short Thread Vials
Supplier: LA PHA PACK
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Canisters, GL 45, b.safe
Supplier: Bohlender
HPLC columns, Purospher® STAR RP-18 endcapped
Supplier: Merck
HPLC columns, ZORBAX® Eclipse XDB
Supplier: Agilent
TLC plates, silica gel 60, unmodified layers, SIL G
Supplier: MACHEREY-NAGEL
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Storage Boxes for Vials
Supplier: MACHEREY-NAGEL
FlashPure EcoFlex Silica Cartridges
Supplier: BUCHI
Screw top vials, pre-assembled Supelco®
Supplier: Merck