Order Entry
Poland
Orders LinkContactUsLinkComponent
Post-mortem scalpel blades
PM60
Catalog # 233-0112
Supplier:   SWANN MORTON
Post-mortem scalpel blades
Catalog # 233-0112
Supplier:  SWANN MORTON
Post-mortem scalpel blades
Post-mortem scalpel blades
PM60
Price
269 zł
Log in for availability
1 * 50 SZT
Product Details & Documents
A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
  • PM40 range with traditional screw fitting
  • PM60 range with standard blade fitment
The PM40B and PM60B blades are 'bull nose' blades for added safety and protection against accidental sharps injuries.
Packaging: All blades are individually wrapped. The PM8 and PM40 stainless steel handles are supplied individually in a protective plastic wallet.
Specifications
  • Type:
    PM60
  • Seq:
    07