Order Entry
Poland
Orders LinkContactUsLinkComponent
Micro suction sets, Aesculap®
Micro suction sets, Aesculap®
Micro suction cannula set, incl. suction handle with finger cut-off and two cannulas each (Ø 0,5 / 0,7 / 1,0 / 1,3 mm), olive Ø 6 - 9 mm, with Luer cone
Catalog # AESCOG970R
Supplier:   Aesculap
Micro suction sets, Aesculap®
Catalog # AESCOG970R
Supplier:  Aesculap
Micro suction sets, Aesculap®
Micro suction cannula set, incl. suction handle with finger cut-off and two cannulas each (Ø 0,5 / 0,7 / 1,0 / 1,3 mm), olive Ø 6 - 9 mm, with Luer cone
Price
Inquire For Price
Log in for availability
1 * 1 SZT
Product Details & Documents
Sets consisting of different micro suction cannulas.
Specifications
  • Description:
    Micro suction cannula set, incl. suction handle with finger cut-off and two cannulas each (Ø 0,5 / 0,7 / 1,0 / 1,3 mm), olive Ø 6 - 9 mm, with Luer cone
  • Seq:
    02