Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Gouges, Aesculap®

Gouge, inclined backward, Wagner, 15 mm, 340 mm
Catalog # AESCND123R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCND123R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge, inclined backward, Wagner, 15 mm, 340 mm

Price
Inquire For Price
Log in for availability
1 * 1 SZT
Product Details & Documents
Specifications
  • Type:
    Wagner
  • Width:
    15 mm
  • Description:
    Gouge, inclined backward
  • Length:
    340 mm
  • Seq:
    44
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.