Order Entry
Poland
ContactUsLinkComponent
INOLAB MULTI 720 BNC,SIMPLE AND RELIABLE BENCH-TOP MULTI-METER (BNC) FOR ROUTINE WORK FOR BATTERY OR MAINS OPERATION. SINGLE INSTRU MENT, WIDE RANGE POWER SUPPLY 1 * 1 items
Catalog # 665-0113
Supplier:  WTW
INOLAB MULTI 720 BNC,SIMPLE AND RELIABLE BENCH-TOP MULTI-METER (BNC) FOR ROUTINE WORK FOR BATTERY OR MAINS OPERATION. SINGLE INSTRU MENT, WIDE RANGE POWER SUPPLY 1 * 1 items
Catalog # 665-0113
Supplier:  WTW
Frequently Bought Together