Order Entry
Poland
Orders LinkContactUsLinkComponent
Hoffmann pinch cocks, heavy type
Hoffmann pinch cocks, heavy type
60 mm
Catalog # BOCH12541
Supplier:   BOCHEM
Hoffmann pinch cocks, heavy type
Catalog # BOCH12541
Supplier:  BOCHEM
Hoffmann pinch cocks, heavy type
Hoffmann pinch cocks, heavy type
60 mm
Price
82,40 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
Hoffmann pinch cocks made of nickel plated brass.
  • Heavy type
  • With flap
Specifications
  • Spanning width:
    60 mm