Order Entry
Poland
Orders LinkContactUsLinkComponent
C Columns, Empty Chromatography Columns
C Columns, Empty Chromatography Columns
C 10/10, Glass, 10×100 mm
Catalog # 19-5001-01
Supplier:   Cytiva
C Columns, Empty Chromatography Columns
Catalog # 19-5001-01
Supplier:  Cytiva
C Columns, Empty Chromatography Columns
C Columns, Empty Chromatography Columns
C 10/10, Glass, 10×100 mm
Price
1 110 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
C columns are economical empty columns suitable for low pressure liquid chromatography and gravity feed at the lower volumes used at research scale.
  • Economical – for low pressure liquid chromatography at research scale
  • Broad compatibility – functions with a stand-alone pump, chromatography system, or gravity feed
  • Precise temperature control – with the optional cooling jacket
  • Efficient packing – packing reservoirs are available as accessories specific to each column diameter
Standard C-series columns have a fixed volume, but they can be upgraded through the addition of flow adapters. These work to keep bed height constant and protected from insoluble particles during chromatography. Adjustable flow adapters are available for 10 mm (AC 10), 16 mm (AC 16), and 26 mm (AC 26) diameter C columns.
Three RC packing reservoirs help optimize packing conditions, especially when packing high beds. They are available for each diameter in the C column series, and are quick and easy to assemble. After the resin is in the column, the RC reservoir can be connected to a pump or Mariotte flask to give optimal flow rates for packing.
Resolution and recovery can often be increased by fitting C columns with a JC thermostatic jacket during temperature-dependent low-pressure chromatography. JC jackets increase control of separation temperature and mechanical resistance of these columns, enabling operating pressures of up to 1 bar.
Delivery information: Each C column comes equipped with polyethylene tubing (1,0 mm inner diameter, 2 m length), bed support screens, snap-on net rings, and instructions for research-scale column chromatography use.
Specifications
  • Description:
    C 10/10
  • Seq:
    01
  • Column hardware:
    Glass
  • Bed dimensions:
    10×100 mm
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Packing reservoirs RC 10
    Packing reservoirs RC 10
    19-5007-01
    Log in for availability
    1 * 1 SZT - 1 170 zł
Product Information
  • Catalog #
    Availability
    Price
  • Support screen for C-10 column
    Support screen for C-10 column
    19-1767-01
    Log in for availability
    1 * 10 SZT - 185 zł
Product Information
  • Catalog #
    Availability
    Price
  • Net rings C 10, AC 10 (10 µm)
    Net rings C 10, AC 10 (10 µm)
    19-5008-01
    Log in for availability
    1 * 5 SZT - 267 zł
Product Information
  • Catalog #
    Availability
    Price
  • Packing reservoirs RC 16
    Packing reservoirs RC 16
    19-5110-01
    Log in for availability
    1 * 1 SZT - 1 130 zł
Product Information
  • Catalog #
    Availability
    Price
  • Tube C 10/40, glass, inner Ø 10 mm
    Tube C 10/40, glass, inner Ø 10 mm
    19-5037-01
    Log in for availability
    1 * 1 SZT - 892 zł
Product Information
  • Catalog #
    Availability
    Price
  • AC 16 flow adapter, for C-16 columns
    AC 16 flow adapter, for C-16 columns
    19-5109-01
    Log in for availability
    1 * 1 SZT - 1 070 zł
Product Information
  • Catalog #
    Availability
    Price
  • Net rings for columns, 16 mm I-Ø, 10 µm
    Net rings for columns, 16 mm I-Ø, 10 µm
    18-8761-01
    Log in for availability
    1 * 5 SZT - 311 zł
Product Information
  • Catalog #
    Availability
    Price
  • O-rings for flow adapters and bottom end pieces, AK 26
    O-rings for flow adapters and bottom end pieces, AK 26
    19-0688-01
    Log in for availability
    1 * 10 SZT - 286 zł
Product Information
  • Catalog #
    Availability
    Price
  • O-rings for flow adapters and bottom end pieces
    O-rings for flow adapters and bottom end pieces
    19-0163-01
    Log in for availability
    1 * 5 SZT - 252 zł
Product Information
  • Catalog #
    Availability
    Price
  • O-rings for flow adapters and bottom end pieces (AC 10)
    O-rings for flow adapters and bottom end pieces (AC 10)
    19-5043-01
    Log in for availability
    1 * 10 SZT - 480 zł
Product Information
  • Catalog #
    Availability
    Price
  • AC 10 flow adapter, for C-10 columns
    AC 10 flow adapter, for C-10 columns
    19-5006-01
    Log in for availability
    1 * 1 SZT - 965 zł
Product Information
  • Catalog #
    Availability
    Price
  • Packing reservoirs RC 26
    Packing reservoirs RC 26
    19-5208-01
    Log in for availability
    1 * 1 SZT - 1 680 zł
Product Information
  • Catalog #
    Availability
    Price
  • Flow adapters AC 26
    Flow adapters AC 26
    19-5207-01
    Log in for availability
    1 * 1 SZT - 1 260 zł