Order Entry
Poland
Orders LinkContactUsLinkComponent
Methyl orange 0,04% in aqueous solution, TECHNICAL pH-indicator

Methyl orange 0,04% in aqueous solution, TECHNICAL pH-indicator

Glass bottle, 1 L
Catalog # 31720.292
Supplier:   VWR Chemicals
Methyl orange 0,04% in aqueous solution, TECHNICAL pH-indicator
Catalog # 31720.292
Supplier:  VWR Chemicals
CAS Number:  547-58-0
Methyl orange 0,04% in aqueous solution, TECHNICAL pH-indicator
Methyl orange 0,04% in aqueous solution, TECHNICAL pH-indicator

Glass bottle, 1 L

CAS Number:  547-58-0
Price
365 zł
Log in for availability
1 * 1 L
Product Details & Documents
  • Transition range: pH 3,1-4,4
Synonyms:  4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
  • CAS Number:547-58-0
  • Formula:C₁₄H₁₄N₃NaO₃S
  • MDL Number:MFCD00007502
  • Molecular Weight:327.34 g/mol
  • Storage Temperature:Ambient
  • EINECS:208-925-3
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.