Order Entry
Poland
Orders LinkContactUsLinkComponent
Sodium hydroxide 50% in aqueous solution, TECHNICAL

Sodium hydroxide 50% in aqueous solution, TECHNICAL

Plastic bottle, 2,5 L
Catalog # 28265.322
Corrosive
Supplier:   VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Catalog # 28265.322
Supplier:  VWR Chemicals
CAS Number:  1310-73-2
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Sodium hydroxide 50% in aqueous solution, TECHNICAL

Plastic bottle, 2,5 L

CAS Number:  1310-73-2
Price
385 zł
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1.53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specification Test Results

  • Assay:
    47 - 51 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L

Specifications