Sodium L(+)-ascorbate ≥99%, TECHNICAL
Plastic bottle for solids, 250 g
Supplier: VWR Chemicals
Sodium L(+)-ascorbate ≥99%, TECHNICAL
Catalog # 27688.235
Supplier: VWR Chemicals
CAS Number: 134-03-2
Jump to:
Product Details & Documents
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Melting Pt:219 °C
- Density:1.799 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-126-1
Documents
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC27688.235 spec_VWRC27688.292
Product specification
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99 %
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



