Order Entry
Poland
Orders LinkContactUsLinkComponent
Mortar spatulas

Mortar spatulas

400 mm, 60 mm, 25 mm
Catalog # BOCH3264
Supplier:   BOCHEM
Mortar spatulas
Catalog # BOCH3264
Supplier:  BOCHEM
Mortar spatulas
Mortar spatulas

400 mm, 60 mm, 25 mm

Price
157 zł
Log in for availability
1 * 1 SZT
Product Details & Documents

Double-ended mortar spatulas made of stainless steel.

Specifications
  • Length:
    400 mm
  • Blade width:
    25 mm
  • Blade length:
    60 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.