Order Entry
Poland
Orders LinkContactUsLinkComponent
Volumetric overflow flasks, without stopper
Volumetric overflow flasks, without stopper
432 ml, Overflow flask
Catalog # NEUB0372-10-432
Volumetric overflow flasks, without stopper
Catalog # NEUB0372-10-432
Volumetric overflow flasks, without stopper
432 ml, Overflow flask
Price
270 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
These volumetric overflow flasks are made of borosilicate glass 3.3.
  • Calibrated to deliver (TD, Ex)
  • Cut and ground exactly at the volume-mark
  • Without stopper
Specially designed for water analysis.
Specifications
  • Capacity:
    432 ml
  • Description:
    Overflow flask
  • Seq:
    07