Taurine
Glass bottle for solids, 100 g
Catalog # BMOL07579-1
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
Product Details & Documents
Synonyms: 2-Aminoethanesulphonic acid
- CAS Number:107-35-7
- Formula:C₂H₇NO₃S
- MDL Number:MFCD00008197
- Molecular Weight:125.15 g/mol
- Merck Index:13,09163
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:203-483-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



