Order Entry
Poland
Orders LinkContactUsLinkComponent
Accessories for TW alpha plus
Accessories for TW alpha plus
Titration head, TZ 1467, for 12 (TZ 1452) and 16 sample (TZ 1459), with 7 drillings NS 14,5, incl. splash shield and rinsing tube
Catalog # SCOI285213671
Supplier:   SI Analytics
Accessories for TW alpha plus
Catalog # SCOI285213671
Supplier:  SI Analytics
Accessories for TW alpha plus
Titration head, TZ 1467, for 12 (TZ 1452) and 16 sample (TZ 1459), with 7 drillings NS 14,5, incl. splash shield and rinsing tube
Price
7 330 zł
Log in for availability
1 * 1 SZT
Product Details & Documents
Specifications
  • Description:
    Titration head, TZ 1467, for 12 (TZ 1452) and 16 sample (TZ 1459), with 7 drillings NS 14,5, incl. splash shield and rinsing tube