Polyaniline, emeraldine base 1 * 5 g
Catalog # 42581.06
Supplier: Thermo Fisher Scientific
Frequently Bought Together
TOLUEN CZDA 1 * 20 L
Supplier: CHEMPUR
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-1,2-Propanediol, pure
Supplier: CHEMPUR
Methanol, pure
Supplier: CHEMPUR
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), pure analytical reagent
Supplier: CHEMPUR
Copper(I) oxide 97%
Supplier: Thermo Fisher Scientific
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific