Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Calcium hypochlorite, tech.

Glass bottle for solids, 50 g
Catalog # 42548.18
Oxidizing
Corrosive
Irritant
Environment
Calcium hypochlorite, tech.
Catalog # 42548.18
CAS Number:  7778-54-3
Calcium hypochlorite, tech.
Calcium hypochlorite, tech.

Glass bottle for solids, 50 g

CAS Number:  7778-54-3
Price
287 zł
Log in for availability
1 * 50 g
Product Details & Documents

CAS No.: 7778-54-3 Molecular Formula: CaCl2O2 Formula Weight: 142.98 Physical Form: Granular Density: 2.350 Appearance: White to pale brown Odour: Like chlorine pH: 11,5 Melting Point: 100° Merck Reference: 14,1674 UN No.: 1748 MDL No.: MFCD00010900

DeOdourant, oxidizing agent, bleaching agent

  • CAS Number:7778-54-3
  • Formula:CaCl₂O₂
  • MDL Number:MFCD00010900
  • Molecular Weight:142.98 g/mol
  • Merck Index:14,1674
  • Melting Pt:100 °C
  • Density:2.350
  • Storage Temperature:Ambient
  • EINECS:231-908-7
  • UN:1748
  • ADR:5.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.