Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(I) oxide ≥99% (metals basis)

Copper(I) oxide ≥99% (metals basis)

2,5 kg
Catalog # 40188.A4
Irritant
Environment
Copper(I) oxide ≥99% (metals basis)
Catalog # 40188.A4
CAS Number:  1317-39-1
Copper(I) oxide ≥99% (metals basis)
Copper(I) oxide ≥99% (metals basis)

2,5 kg

CAS Number:  1317-39-1
Price
7 320 zł
Log in for availability
1 * 2,5 kg
Product Details & Documents

Purity: 99% CAS No.: 1317-39-1 Molecular Formula: Cu2O Formula Weight: 143.08 Physical Form: -200 Mesh Powder Density: 6.0 Refractive Index: 2.705 Sensitivity: Air and Moisture Sensitive Appearance: Red to dark red Odour: Odourless Melting Point: 1235° Boiling Point: 1800° dec. Merck Reference: 14,2664 UN No.: 3077 MDL No.: MFCD00010974

Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143.09 g/mol
  • Merck Index:14,2664
  • Boiling Pt:1800 °C (dec.)
  • Melting Pt:1235 °C
  • Density:6.0
  • Storage Temperature:Ambient
  • EINECS:215-270-7
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    2,5 kg

Specifications