Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Copper(II) carbonate basic (≥55% Cu)

2 kg
Catalog # 33305.A3
Irritant
Copper(II) carbonate basic (≥55% Cu)
Catalog # 33305.A3
CAS Number:  12069-69-1
Copper(II) carbonate basic (≥55% Cu)
Copper(II) carbonate basic (≥55% Cu)

2 kg

CAS Number:  12069-69-1
Price
1 750 zł
Log in for availability
1 * 2 kg
Product Details & Documents

CAS No.: 12069-69-1 Molecular Formula: CH2Cu2O5 Formula Weight: 221.11 Physical Form: Powder Density: 4.00 Appearance: Green Melting Point: 200° dec. Merck Reference: 14,2631 MDL No.: MFCD00010976

Synonyms:  Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
  • CAS Number:12069-69-1
  • Formula:CuCO₃·Cu(OH)₂
  • MDL Number:MFCD00010976
  • Molecular Weight:221.12 g/mol
  • Merck Index:14,2631
  • Melting Pt:200 °C (dec.)
  • Density:4.00
  • Storage Temperature:Ambient
  • EINECS:235-113-6
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    2 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.