Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(II) carbonate basic (≥55% Cu)

Copper(II) carbonate basic (≥55% Cu)

Glass bottle for solids, 500 g
Catalog # 33305.36
Irritant
Copper(II) carbonate basic (≥55% Cu)
Catalog # 33305.36
CAS Number:  12069-69-1
Copper(II) carbonate basic (≥55% Cu)
Copper(II) carbonate basic (≥55% Cu)

Glass bottle for solids, 500 g

CAS Number:  12069-69-1
Price
545 zł
Log in for availability
1 * 500 g
Product Details & Documents

CAS No.: 12069-69-1 Molecular Formula: CH2Cu2O5 Formula Weight: 221.11 Physical Form: Powder Density: 4.00 Appearance: Green Melting Point: 200° dec. Merck Reference: 14,2631 MDL No.: MFCD00010976

Synonyms:  Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
  • CAS Number:12069-69-1
  • Formula:CuCO₃·Cu(OH)₂
  • MDL Number:MFCD00010976
  • Molecular Weight:221.12 g/mol
  • Merck Index:14,2631
  • Melting Pt:200 °C (dec.)
  • Density:4.00
  • Storage Temperature:Ambient
  • EINECS:235-113-6
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications