penta-Sodium triphosphate
Plastic bottle for solids, 2 kg
Supplier: Thermo Fisher Scientific
penta-Sodium triphosphate
Catalog # 13440.A3
Supplier: Thermo Fisher Scientific
CAS Number: 7758-29-4
Jump to:
Product Details & Documents
CAS No.: 7758-29-4 Molecular Formula: Na5P3O10 Formula Weight: 367.91 Physical Form: Granular Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 622° Solubility: Soluble in water Merck Reference: 14,8697 MDL No.: MFCD00003514
Synonyms: Sodium triphosphate pentabasic, Sodium tripolyphosphate
- CAS Number:7758-29-4
- Formula:Na₅O₁₀P₃
- MDL Number:MFCD00003514
- Molecular Weight:367.86 g/mol
- Merck Index:14,8697
- Melting Pt:622 °C
- Density:2.52 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-838-7
Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:2 kg
- Cat. No.:13440.A3



