penta-Sodium triphosphate
Plastic bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
penta-Sodium triphosphate
Catalog # 13440.36
Supplier: Thermo Fisher Scientific
CAS Number: 7758-29-4
Jump to:
Product Details & Documents
CAS No.: 7758-29-4 Molecular Formula: Na5P3O10 Formula Weight: 367.91 Physical Form: Granular Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 622° Solubility: Soluble in water Merck Reference: 14,8697 MDL No.: MFCD00003514
Synonyms: Sodium triphosphate pentabasic, Sodium tripolyphosphate
- CAS Number:7758-29-4
- Formula:Na₅O₁₀P₃
- MDL Number:MFCD00003514
- Molecular Weight:367.86 g/mol
- Merck Index:14,8697
- Melting Pt:622 °C
- Density:2.52 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-838-7
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



