Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(I) oxide ≥97% (Cu + Cu₂O)

Copper(I) oxide ≥97% (Cu + Cu₂O)

2 kg
Catalog # 12300.A3
Irritant
Environment
Copper(I) oxide ≥97% (Cu + Cu₂O)
Catalog # 12300.A3
CAS Number:  1317-39-1
Copper(I) oxide ≥97% (Cu + Cu₂O)
Copper(I) oxide ≥97% (Cu + Cu₂O)

2 kg

CAS Number:  1317-39-1
Price
878 zł
Log in for availability
1 * 2 kg
Product Details & Documents

Purity: 97% CAS No.: 1317-39-1 Molecular Formula: Cu2O Formula Weight: 143.08 Physical Form: -325 Mesh Powder Density: 6.0 Refractive Index: 2.705 Sensitivity: Air and Moisture Sensitive Appearance: Purple-red Odour: Odourless Melting Point: 1235° Boiling Point: 1800° dec. Solubility: Practically insoluble in water. Soluble in NH4OH Merck Reference: 14,2664 UN No.: 3077 MDL No.: MFCD00010974

Red pigment for glass and ceramic glazes, and as a catalyst

Synonyms:  Cuprous oxide, Dicopper oxide
  • CAS Number:1317-39-1
  • Formula:Cu₂O
  • MDL Number:MFCD00010974
  • Molecular Weight:143.09 g/mol
  • Merck Index:14,2664
  • Boiling Pt:1800 °C (dec.)
  • Melting Pt:1235 °C
  • Density:6.0
  • Storage Temperature:Ambient
  • EINECS:215-270-7
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    2 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.