Order Entry
Poland
Orders LinkContactUsLinkComponent
Pharmalytes® IEF carrier ampholytes

Pharmalytes® IEF carrier ampholytes

Pharmalyte®, broad range pH 5 - 8
Catalog # 17-0453-01
Supplier:   Cytiva
Pharmalytes® IEF carrier ampholytes
Catalog # 17-0453-01
Supplier:  Cytiva
CAS Number:  
Pharmalytes® IEF carrier ampholytes
Pharmalytes® IEF carrier ampholytes

Pharmalyte®, broad range pH 5 - 8

CAS Number:  
Price
4 450 zł
Log in for availability
1 * 25 mL
Product Details & Documents

Pharmalyte® carrier ampholytes, prepared by the co-polymerization of glycine, glycylglycine, amines and epichlorohydrin, are available in five broad-range and four narrow-range pH intervals.

  • Carrier for high resolution isoelectric focusing (IEF)
  • Soluble in cold water and hot water
  • Stored at 2 to 8 °C

Pharmalyte® broad range can be used directly for agarose, Ultrodex or polyacrylamide gels and form linear gradients. Pharmalyte narrow range are intervals for special dedicated applications in polyacrylamide gels.

Each interval contains numerous ampholytes with a high buffering capacity per pH unit. Pharmalytes® carrier ampholytes form an extremely stable linear pH gradient and exhibit even conductivity across the gel.

Specifications

Specifications

  • Description:
    Pharmalyte®, broad range pH 5 - 8

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.