Order Entry
Poland
Orders LinkContactUsLinkComponent
Post-mortem scalpel blades | Avantor
Post-mortem scalpel blades
PM40
numer katalogowy 233-0110
Dostawca:   SWANN MORTON
Post-mortem scalpel blades
numer katalogowy 233-0110
Dostawca:  SWANN MORTON
Post-mortem scalpel blades
Post-mortem scalpel blades
PM40
Przejdź do:
Price
189 zł
Zaloguj się dla dostępności
1 * 10 SZT
Product Details & Documents
A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
  • PM40 range with traditional screw fitting
  • PM60 range with standard blade fitment
The PM40B and PM60B blades are 'bull nose' blades for added safety and protection against accidental sharps injuries.
Sposób pakowania: All blades are individually wrapped. The PM8 and PM40 stainless steel handles are supplied individually in a protective plastic wallet.
Dane techniczne
  • Type:
    PM40
  • Seq:
    05