Test kits, TOC, Spectroquant®, Supelco®
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*5 TOC screw caps for digestion required.
Documents
- Chemical method:Indicator
- Pk:25 Tests
- Description:TOC cell test*<sup>5</sup>
- For unit:Prove, Nova 30/60, Multy
- Range:5,0 - 80,0 mg/l TOC
- Cat. No.:1.14878.0001
- M-Clarity quality level:MQ100