Cyclohexanone, EMPLURA®, extra pure, Supelco®
Catalog # 1.02888.9191
Supplier: Merck
CAS Number: 108-94-1
Jump to:
Product Details & Documents
Synonyms: Pimelic Ketone
- CAS Number:108-94-1
- Formula:C₆H₁₀(=O)
- MDL Number:MFCD00001625
- Molecular Weight:98.14 g/mol
- Merck Index:13,02755
- Boiling Pt:155.6 °C (1013 hPa)
- Melting Pt:–31 °C
- Density:0.9478 g/cm³ (20 °C)
- Flash Pt:43 °C
- UN:1915
- ADR:3,III
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:203-631-1
- REACH:01-2119453616-35
Documents
Certificates
Search by Lot NumberMERC1.02888.1000_TDS_EN MERC1.02888.2500_TDS_EN ProductPage_102888
Product specification
msds_102888
SDS
Specifications
Specifications
- Pack type:Steel barrel
- Pk:190 L
- Cat. No.:1.02888.9191
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



