Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Sealing caps, Nalgene®

24 mm, 50 ml
Catalog # 525-2924
   Low Manufacturing Impact
Sealing caps, Nalgene®
Catalog # 525-2924
   Low Manufacturing Impact
Sealing caps, Nalgene®
Sealing caps, Nalgene®

24 mm, 50 ml

Price
172 zł
Log in for availability
1 * 2 SZT
Product Details & Documents

PP, autoclavable.

  • Silicone gasket

For use at recommended maximum speeds 10000×g without leakage, even when full.

Specifications
  • For Ø ext.:
    24 mm
  • For tubes:
    50 ml
  • Seq:
    02
  • Cat. No.:
    525-2924
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact