Sealing cap assemblies, Nalgene®
38 mm, 250 ml
Catalog # 525-2915
Supplier: Thermo Fisher Scientific
Sealing cap assemblies, Nalgene®
Catalog # 525-2915
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Tefzel™ FEP plug, PP screw closure, Viton™ fluoroelastomer O-ring.
- PP screw closure, ETFE plug, Viton™ fluoroelastomer O-ring
- Top fits securely for use at maximum speeds without leakage
- Autoclavable/biohazard/leakproof
Process tubes at their highest rated speed level (10000×g or greater) without leakage, even when full. Sealing cap assemblies include a Viton™ fluoroelastomer O-ring. Also recommended when spinning hazardous materials compatible with centrifuge tube material.
Specifications
- For Ø ext.:38 mm
- For tubes:250 ml
- Seq:03
- Cat. No.:525-2915
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



